1-(Cyclooctylamino)-6-methyl-9H-xanthen-9-one ≥95%

Supplier: Matrix Scientific

045703-500MG 045703-1G 045703-5G
101872-186EA 375.38 USD
101872-186 101872-188 101872-190
1-(Cyclooctylamino)-6-methyl-9H-xanthen-9-one ≥95%
1-(Cyclooctylamino)-6-methyl-9H-xanthen-9-one

Hazard: Irritant

Formula: C₂₂H₂₅NO₂
Melting Pt: 126…129 °C
MDL Number: MFCD03425817
CAS Number: 886360-53-8

Order Now

Specification Test Results

Melting point 126-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR