Ethyl-5-(aminocarbonyl)-4-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

045701-500MG 045701-1G
101872-176EA 278.46 USD
101872-176 101872-178
Ethyl-5-(aminocarbonyl)-4-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%
Ethyl-5-(aminocarbonyl)-4-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
MW: 224.22 g/mol
Melting Pt: 300…303 °C
MDL Number: MFCD00973779
CAS Number: 171113-29-4

Order Now

Specification Test Results

Melting point 300-303°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR