3-(1,3-Benzoxazol-2-yl)-4-fluoroaniline ≥95%
Supplier: Matrix Scientific
045698-500MG
045698-1G
101872-166EA
492.31
USD
101872-166
101872-168
3-(1,3-Benzoxazol-2-yl)-4-fluoroaniline ≥95%
3-(1,3-Benzoxazol-2-yl)-4-fluoroaniline
Hazard: Irritant
Formula:
C₁₃H₉FN₂O Melting Pt: 153…156 °C |
MDL Number:
MFCD03425815 CAS Number: 220705-28-2 |
Specification Test Results
Melting point | 153-156°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...