2-(4-Aminophenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045696-500MG 045696-1G
101872-156EA 200 USD
101872-156 101872-158
2-(4-Aminophenyl)propanoic acid ≥95%
2-(4-Aminophenyl)propanoic acid

Hazard: Irritant

Formula: C₉H₁₁NO₂
Melting Pt: 132…134 °C
MDL Number: MFCD00800233
CAS Number: 59430-62-5

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR