5-[(3-Nitrophenyl)methylene]-1,3-thiazolane-2,4-dione ≥95%

Supplier: Matrix Scientific

045694-500MG 045694-1G 045694-5G
101872-144EA 181.54 USD
101872-144 101872-146 101872-148
5-[(3-Nitrophenyl)methylene]-1,3-thiazolane-2,4-dione ≥95%
5-[(3-Nitrophenyl)methylene]-1,3-thiazolane-2,4-dione

Hazard: Irritant

Formula: C₁₀H₆N₂O₄S
MW: 250.23 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD00817881
CAS Number: 24044-52-8

Order Now

Specification Test Results

Melting point 185-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR