3-Amino-3-(3-cyclohexenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045691-500MG 045691-1G 045691-5G
101872-126EA 278.46 USD
101872-126 101872-128 101872-130
3-Amino-3-(3-cyclohexenyl)propanoic acid ≥95%
3-Amino-3-(3-cyclohexenyl)propanoic acid

Hazard: Irritant

Formula: C₉H₁₅NO₂
Melting Pt: 209…211 °C
MDL Number: MFCD02653301
CAS Number: 40125-69-7

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR