Dimethyl-2-(4-aminophenyl)malonate ≥97%

Supplier: Matrix Scientific

045688-500MG 045688-1G 045688-5G
101872-114EA 178.46 USD
101872-114 101872-116 101872-118
Dimethyl-2-(4-aminophenyl)malonate ≥97%
Dimethyl-2-(4-aminophenyl)malonate

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD03305797
CAS Number: 183473-82-7

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR