3-[4-(tert-Butyl)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-tert-Butylcinnamic acid

045683-500MG 045683-1G 045683-5G
101872-090EA 41.54 USD
101872-090 101872-092 101872-094
3-[4-(tert-Butyl)phenyl]acrylic acid ≥95%
3-[4-(tert-Butyl)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Melting Pt: 195…197 °C
MDL Number: MFCD00040719
CAS Number: 1208-65-7

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR