3-Amino-3-[4-(tert-butyl)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045681-500MG 045681-1G 045681-5G
101872-078EA 124.62 USD
101872-078 101872-080 101872-082
3-Amino-3-[4-(tert-butyl)phenyl]propanoic acid ≥95%
3-Amino-3-[4-(tert-butyl)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
Melting Pt: 240…242 °C
MDL Number: MFCD02153019
CAS Number: 282524-82-7

Order Now

Specification Test Results

Melting point 240-242°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR