3-Amino-3-(3-thienyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045680-500MG 045680-1G 045680-5G
101872-072EA 140 USD
101872-072 101872-074 101872-076
3-Amino-3-(3-thienyl)propanoic acid ≥95%
3-Amino-3-(3-thienyl)propanoic acid

Hazard: Irritant

Formula: C₇H₉NO₂S
Melting Pt: 208…210 °C
MDL Number: MFCD00152179
CAS Number: 94333-62-7

Order Now

Specification Test Results

Melting point 208-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR