3-(Boc-amino)-3-(3-methylphenyl)propionic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-3-(m-tolyl)propanoic acid
045678-500MG
045678-1G
101872-068EA
595.38
USD
101872-068
101872-070
3-(Boc-amino)-3-(3-methylphenyl)propionic acid ≥95%
3-(Boc-amino)-3-(3-methylphenyl)propionic acid
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄ MW: 279.3315 g/mol Melting Pt: 96-98°C |
MDL Number:
MFCD02090699 CAS Number: 284493-57-8 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...