3-(Boc-amino)-3-(2-methylphenyl)propionic acid ≥95%

Supplier: Matrix Scientific

045677-500MG 045677-1G
101872-064EA 515.38 USD
101872-064 101872-066
3-(Boc-amino)-3-(2-methylphenyl)propionic acid ≥95%
3-(Boc-amino)-3-(2-methylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
Melting Pt: 137…139 °C
MDL Number: MFCD02090696
CAS Number: 284493-54-5

Order Now

Specification Test Results

Melting point 137-139°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR