3-Amino-3-(3-nitrophenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-3-(3-nitrophenyl)propanoic acid

045676-500MG 045676-1G 045676-5G
101872-058EA 278.46 USD
101872-058 101872-060 101872-062
3-Amino-3-(3-nitrophenyl)propionic acid ≥95%
3-Amino-3-(3-nitrophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₄
MW: 210.1867 g/mol
Melting Pt: 213-215°C
MDL Number: MFCD00472612
CAS Number: 5678-47-7

Order Now

Specification Test Results

Melting point 213-215°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR