3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

045674-500MG 045674-1G
101872-050EA 556.92 USD
101872-050 101872-052
3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid ≥95%
3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 295.34 g/mol
Melting Pt: 77…79 °C
MDL Number: MFCD02090693
CAS Number: 284493-53-4

Order Now

Specification Test Results

Melting point 77-79°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR