4-(2-Thienyl)-2-pyrimidinamine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-4-(thien-2-yl)pyrimidine
045673-500MG
045673-1G
045673-5G
101872-044EA
375.38
USD
101872-044
101872-046
101872-048
4-(2-Thienyl)-2-pyrimidinamine ≥95%
4-(2-Thienyl)-2-pyrimidinamine
Hazard: Irritant
Formula:
C₈H₇N₃S MW: 177.23 g/mol Melting Pt: 187…190 °C |
MDL Number:
MFCD03001297 CAS Number: 154321-60-5 |
Specification Test Results
Melting point | 187-190°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...