2-[1-(2-Furyl)ethylidene]malononitrile ≥95%
Supplier: Matrix Scientific
045670-500MG
045670-1G
045670-5G
101872-026EA
181.54
USD
101872-026
101872-028
101872-030
2-[1-(2-Furyl)ethylidene]malononitrile ≥95%
2-[1-(2-Furyl)ethylidene]malononitrile
Hazard: Irritant
Formula:
C₉H₆N₂O Melting Pt: 86…89 °C |
MDL Number:
MFCD01570319 CAS Number: 62737-71-7 |
Specification Test Results
Melting point | 86-89°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...