1-Oxo-3-phenyl-3,4-dihydro-1H-isochromene-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

045668-500MG 045668-1G
101872-016EA 323.08 USD
101872-016 101872-018
1-Oxo-3-phenyl-3,4-dihydro-1H-isochromene-4-carboxylic acid ≥95%
1-Oxo-3-phenyl-3,4-dihydro-1H-isochromene-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₂O₄
Melting Pt: 185…187 °C
MDL Number: MFCD01845966
CAS Number: 68204-74-0

Order Now

Specification Test Results

Melting point 185-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR