2-[1-(2-Thienyl)ethylidene]malononitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-(Thiophen-2-yl)ethylidene)malononitrile

045667-500MG 045667-1G 045667-5G
101872-010EA 181.54 USD
101872-010 101872-012 101872-014
2-[1-(2-Thienyl)ethylidene]malononitrile ≥95%
2-[1-(2-Thienyl)ethylidene]malononitrile

Hazard: Irritant

Formula: C₉H₆N₂S
MW: 174.23 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD00184599
CAS Number: 10432-44-7

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR