1-[5-(2,2,2-Trichloroacetyl)-1H-pyrrol-3-yl]-1-butanone ≥95%

Supplier: Matrix Scientific

045664-500MG 045664-1G
101871-994EA 556.92 USD
101871-994 101871-996
1-[5-(2,2,2-Trichloroacetyl)-1H-pyrrol-3-yl]-1-butanone ≥95%
1-[5-(2,2,2-Trichloroacetyl)-1H-pyrrol-3-yl]-1-butanone

Hazard: Irritant

Formula: C₁₀H₁₀Cl₃NO₂
Melting Pt: 133…135 °C
MDL Number: MFCD03001293
CAS Number: 111468-91-8

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR