Methyl-4-(2,4-dichlorobenzoyl)-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

045661-500MG 045661-1G
101871-982EA 747.69 USD
101871-982 101871-984
Methyl-4-(2,4-dichlorobenzoyl)-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-(2,4-dichlorobenzoyl)-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₉Cl₂NO₃
Melting Pt: 172…175 °C
MDL Number: MFCD03001291
CAS Number: 453557-85-2

Order Now

Specification Test Results

Melting point 172-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR