2,2,2-Trichloro-1-[4-(4-methoxybenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

045660-500MG 045660-1G
101871-978EA 556.92 USD
101871-978 101871-980
2,2,2-Trichloro-1-[4-(4-methoxybenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%
2,2,2-Trichloro-1-[4-(4-methoxybenzoyl)-1H-pyrrol-2-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₄H₁₀Cl₃NO₃
Melting Pt: 146…149 °C
MDL Number: MFCD03001290
CAS Number: 174417-17-5

Order Now

Specification Test Results

Melting point 146-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR