5-[(Dimethylamino)methylene]-3-ethyl-2-(4H-1,2,4-triazol-4-ylamino)-1,3-thiazolan-4-one ≥95%
Supplier: Matrix Scientific
045658-500MG
045658-1G
101871-970EA
375.38
USD
101871-970
101871-972
5-[(Dimethylamino)methylene]-3-ethyl-2-(4H-1,2,4-triazol-4-ylamino)-1,3-thiazolan-4-one ≥95%
5-[(Dimethylamino)methylene]-3-ethyl-2-(4H-1,2,4-triazol-4-ylamino)-1,3-thiazolan-4-one
Hazard: Irritant
Formula:
C₁₀H₁₆N₆OS MW: 268.34 g/mol Melting Pt: 266.32 °C |
MDL Number:
MFCD05021865 CAS Number: 453557-84-1 |
Specification Test Results
Melting point | 266.32°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...