Ethyl-4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzoate

045656-500MG 045656-1G
101871-960EA 375.38 USD
101871-960 101871-962
Ethyl-4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate ≥95%
Ethyl-4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₇NO₃
MW: 271.32 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD00022461
CAS Number: 52034-37-4

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR