Ethyl-4-(2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
045655-500MG
045655-1G
045655-5G
101871-954EA
181.54
USD
101871-954
101871-956
101871-958
Ethyl-4-(2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate ≥95%
Ethyl-4-(2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₇NO₂ Melting Pt: 90…92 °C |
MDL Number:
MFCD00022462 CAS Number: 5159-70-6 |
Specification Test Results
Melting point | 90-92°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...