2-(2-Formyl-6-methoxyphenoxy)acetonitrile ≥95%

Supplier: Matrix Scientific

045650-500MG 045650-1G 045650-5G
101871-930EA 75.38 USD
101871-930 101871-932 101871-934
2-(2-Formyl-6-methoxyphenoxy)acetonitrile ≥95%
2-(2-Formyl-6-methoxyphenoxy)acetonitrile

Hazard: Irritant

Formula: C₁₀H₉NO₃
Melting Pt: 117…119 °C
MDL Number: MFCD02082429
CAS Number: 127500-89-4

Order Now

Specification Test Results

Melting point 114-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR