1,1,1-Trifluoro-3-{4-[3-(trifluoromethyl)phenyl]piperazino}-2-propanol ≥95%

Supplier: Matrix Scientific

045649-500MG 045649-1G
101871-926EA 1296.92 USD
101871-926 101871-928
1,1,1-Trifluoro-3-{4-[3-(trifluoromethyl)phenyl]piperazino}-2-propanol ≥95%
1,1,1-Trifluoro-3-{4-[3-(trifluoromethyl)phenyl]piperazino}-2-propanol

Hazard: Irritant

Formula: C₁₄H₁₆F₆N₂O
Melting Pt: 68…71 °C
MDL Number: MFCD01871732
CAS Number: 453557-82-9

Order Now

Specification Test Results

Melting point 68-71°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR