1,1,1-Trifluoro-3-[4-(2-fluorophenyl)piperazino]-2-propanol ≥95%
Supplier: Matrix Scientific
045646-500MG
045646-1G
101871-910EA
1296.92
USD
101871-910
101871-912
1,1,1-Trifluoro-3-[4-(2-fluorophenyl)piperazino]-2-propanol ≥95%
1,1,1-Trifluoro-3-[4-(2-fluorophenyl)piperazino]-2-propanol
Hazard: Irritant
Formula:
C₁₃H₁₆F₄N₂O Melting Pt: 92…95 °C |
MDL Number:
MFCD01871727 CAS Number: 453557-80-7 |
Specification Test Results
Melting point | 92-95°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...