2-(2-Formyl-4-methoxyphenoxy)acetonitrile ≥95%

Supplier: Matrix Scientific

045643-500MG 045643-1G
101871-900EA 278.46 USD
101871-900 101871-902
2-(2-Formyl-4-methoxyphenoxy)acetonitrile ≥95%
2-(2-Formyl-4-methoxyphenoxy)acetonitrile

Hazard: Irritant

Formula: C₁₀H₉NO₃
Melting Pt: 68…69 °C
MDL Number: MFCD02082011
CAS Number: 149384-09-8

Order Now

Specification Test Results

Melting point 68-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR