1,1,1-Trifluoro-3-{[2-(trifluoromethyl)benzyl]amino}-2-propanol ≥95%

Supplier: Matrix Scientific

045642-500MG 045642-1G
101871-896EA 1109.23 USD
101871-896 101871-898
1,1,1-Trifluoro-3-{[2-(trifluoromethyl)benzyl]amino}-2-propanol ≥95%
1,1,1-Trifluoro-3-{[2-(trifluoromethyl)benzyl]amino}-2-propanol

Hazard: Irritant

Formula: C₁₁H₁₁F₆NO
MW: 287.21 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD01871716
CAS Number: 453557-79-4

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR