2-Chloro-5-(1H-pyrazol-1-ylmethyl)-1,3-thiazole ≥95%

Supplier: Matrix Scientific

045641-500MG 045641-1G 045641-5G
101871-890EA 278.46 USD
101871-890 101871-892 101871-894
2-Chloro-5-(1H-pyrazol-1-ylmethyl)-1,3-thiazole ≥95%
2-Chloro-5-(1H-pyrazol-1-ylmethyl)-1,3-thiazole

Hazard: Irritant

Formula: C₇H₆ClN₃S
MW: 199.66 g/mol
Melting Pt: 50…52 °C
MDL Number: MFCD01871711
CAS Number: 453557-78-3

Order Now

Specification Test Results

Melting point 50-52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR