1H-Pyrrole-2-carbaldehyde oxime ≥95%

Supplier: Matrix Scientific

045640-500MG 045640-1G 045640-5G
101871-884EA 181.54 USD
101871-884 101871-886 101871-888
1H-Pyrrole-2-carbaldehyde oxime ≥95%
1H-Pyrrole-2-carbaldehyde oxime

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD00965095
CAS Number: 32597-34-5

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR