Methyl-3-(benzoylamino)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045634-500MG 045634-1G 045634-5G
101871-856EA 181.54 USD
101871-856 101871-858 101871-860
Methyl-3-(benzoylamino)-2-thiophenecarboxylate ≥95%
Methyl-3-(benzoylamino)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁NO₃S
Melting Pt: 99…101 °C
MDL Number: MFCD00665577
CAS Number: 79128-70-4

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR