2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile ≥95%
Supplier: Matrix Scientific
045633-500MG
045633-1G
045633-5G
101871-850EA
375.38
USD
101871-850
101871-852
101871-854
2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile ≥95%
2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile
Hazard: Irritant
Formula:
C₁₂H₉N₃O MW: 211.23 g/mol Melting Pt: 85…87 °C |
MDL Number:
MFCD01443818 CAS Number: 339105-55-4 |
Specification Test Results
Melting point | 85-87°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...