Methyl-4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

045631-500MG 045631-1G 045631-5G
101871-838EA 375.38 USD
101871-838 101871-840 101871-842
Methyl-4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate ≥95%
Methyl-4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₃S
Melting Pt: 108…109 °C
MDL Number: MFCD01568814
CAS Number: 339105-21-4

Order Now

Specification Test Results

Melting point 108-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR