4-[(2-Chloro-1,3-thiazol-5-yl)methyl]morpholine ≥95%

Supplier: Matrix Scientific

045630-500MG 045630-1G 045630-5G
101871-832EA 93.85 USD
101871-832 101871-834 101871-836
4-[(2-Chloro-1,3-thiazol-5-yl)methyl]morpholine ≥95%
4-[(2-Chloro-1,3-thiazol-5-yl)methyl]morpholine

Hazard: Irritant

Formula: C₈H₁₁ClN₂OS
Melting Pt: 31…34 °C
MDL Number: MFCD01443905
CAS Number: 339105-10-1

Order Now

Specification Test Results

Melting point 31-34°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR