2-[4-(1H-Pyrazol-3-yl)phenoxy]pyrimidine ≥95%

Supplier: Matrix Scientific

045629-500MG 045629-1G
101871-828EA 556.92 USD
101871-828 101871-830
2-[4-(1H-Pyrazol-3-yl)phenoxy]pyrimidine ≥95%
2-[4-(1H-Pyrazol-3-yl)phenoxy]pyrimidine

Hazard: Irritant

Formula: C₁₃H₁₀N₄O
Melting Pt: 139…141 °C
MDL Number: MFCD01443797
CAS Number: 321998-82-7

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR