1-{4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

045628-500MG 045628-1G 045628-5G
101871-822EA 375.38 USD
101871-822 101871-824 101871-826
1-{4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}-1-ethanone ≥95%
1-{4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂S
Melting Pt: 85…86 °C
MDL Number: MFCD01568813
CAS Number: 339104-75-5

Order Now

Specification Test Results

Melting point 85-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR