1-{3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

045626-500MG 045626-1G 045626-5G
101871-810EA 375.38 USD
101871-810 101871-812 101871-814
1-{3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}-1-ethanone ≥95%
1-{3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂S
Melting Pt: 80…82 °C
MDL Number: MFCD01568812
CAS Number: 339104-67-5

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR