Ethyl-2-(2,4,5-trioxo-1-imidazolidinyl)acetate ≥95%

Supplier: Matrix Scientific

045623-500MG 045623-1G 045623-5G
101871-792EA 278.46 USD
101871-792 101871-794 101871-796
Ethyl-2-(2,4,5-trioxo-1-imidazolidinyl)acetate ≥95%
Ethyl-2-(2,4,5-trioxo-1-imidazolidinyl)acetate

Hazard: Irritant

Formula: C₇H₈N₂O₅
MW: 200.15 g/mol
Melting Pt: 160…162 °C
MDL Number: MFCD00665817
CAS Number: 89694-35-9

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR