3-(4-Bromophenyl)-5-methyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

045620-500MG 045620-1G 045620-5G
101871-774EA 253.85 USD
101871-774 101871-776 101871-778
3-(4-Bromophenyl)-5-methyl-1H-pyrazole ≥95%
3-(4-Bromophenyl)-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₉BrN₂
Melting Pt: 163…165 °C
MDL Number: MFCD02083151
CAS Number: 145353-53-3

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR