2-Chloro-4-phenylnicotinonitrile ≥95%

Supplier: Matrix Scientific

045617-500MG 045617-1G 045617-5G
101871-768EA 556.92 USD
101871-768 101871-770 101871-772
2-Chloro-4-phenylnicotinonitrile ≥95%
2-Chloro-4-phenylnicotinonitrile

Hazard: Irritant

Formula: C₁₂H₇ClN₂
Melting Pt: 125…127.5 °C
MDL Number: MFCD01935958
CAS Number: 163563-64-2

Order Now

Specification Test Results

Melting point 125-127.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR