Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate ≥95%

Supplier: Matrix Scientific

045615-500MG 045615-1G 045615-5G
101871-762EA 747.69 USD
101871-762 101871-764 101871-766
Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate ≥95%
Methyl-4-[2-amino-4-(trifluoromethyl)anilino]butanoate

Hazard: Irritant

Formula: C₁₂H₁₅F₃N₂O₂
MW: 276.26 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD00793102
CAS Number: 339101-34-7

Order Now

Specification Test Results

Melting point 91-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR