4-[(3-Nitro-2-pyridinyl)amino]butanoic acid ≥95%

Supplier: Matrix Scientific

045614-500MG 045614-1G 045614-5G
101871-756EA 747.69 USD
101871-756 101871-758 101871-760
4-[(3-Nitro-2-pyridinyl)amino]butanoic acid ≥95%
4-[(3-Nitro-2-pyridinyl)amino]butanoic acid

Hazard: Irritant

Formula: C₉H₁₁N₃O₄
Melting Pt: 98…100 °C
MDL Number: MFCD00793101
CAS Number: 339101-31-4

Order Now

Specification Test Results

Melting point 98-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR