Methyl-4-[2-nitro-4-(trifluoromethyl)anilino]butanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-[2-nitro-4-(trifluoromethyl)anilino]butyrate

045613-500MG 045613-1G 045613-5G
101871-750EA 181.54 USD
101871-750 101871-752 101871-754
Methyl-4-[2-nitro-4-(trifluoromethyl)anilino]butanoate ≥95%
Methyl-4-[2-nitro-4-(trifluoromethyl)anilino]butanoate

Hazard: Irritant

Formula: C₁₂H₁₃F₃N₂O₄
MW: 306.24 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD00793100
CAS Number: 339101-25-6

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR