Methyl-2-[(3-chloro-2,2-dimethylpropanoyl)amino]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(3-chloro-2,2-dimethylpropanamido)benzoate

045610-500MG 045610-1G 045610-5G
101871-734EA 181.54 USD
101871-734 101871-736 101871-738
Methyl-2-[(3-chloro-2,2-dimethylpropanoyl)amino]benzenecarboxylate ≥95%
Methyl-2-[(3-chloro-2,2-dimethylpropanoyl)amino]benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₃
MW: 269.73 g/mol
Melting Pt: 53…55 °C
MDL Number: MFCD00793430
CAS Number: 339100-86-6

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR