Methyl-3-(3,3-dimethyl-2-oxo-1-azetanyl)-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
045609-500MG
045609-1G
045609-5G
101871-728EA
747.69
USD
101871-728
101871-730
101871-732
Methyl-3-(3,3-dimethyl-2-oxo-1-azetanyl)-2-thiophenecarboxylate ≥95%
Methyl-3-(3,3-dimethyl-2-oxo-1-azetanyl)-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₃NO₃S Melting Pt: 94…96 °C |
MDL Number:
MFCD00793418 CAS Number: 339100-30-0 |
Specification Test Results
Melting point | 94-96°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...