Methyl-4-[3-(4-piperidinyl)-1H-pyrazol-5-yl]phenyl ether ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[5-(4-Methoxyphenyl)-1H-pyrazol-3-yl]piperidine

045608-500MG 045608-1G
101871-724EA 747.69 USD
101871-724 101871-726
Methyl-4-[3-(4-piperidinyl)-1H-pyrazol-5-yl]phenyl ether ≥95%
Methyl-4-[3-(4-piperidinyl)-1H-pyrazol-5-yl]phenyl ether

Hazard: Irritant

Formula: C₁₅H₁₉N₃O
MW: 257.34 g/mol
Melting Pt: 181…183 °C
MDL Number: MFCD00665098
CAS Number: 103660-47-5

Order Now

Specification Test Results

Melting point 181-183°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR