2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-phenylpropanoic acid ≥95%
Supplier: Matrix Scientific
045603-500MG
045603-1G
045603-5G
101871-700EA
181.54
USD
101871-700
101871-702
101871-704
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-phenylpropanoic acid ≥95%
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-phenylpropanoic acid
Hazard: Irritant
Formula:
C₁₇H₁₅NO₃ MW: 281.31 g/mol Melting Pt: 212…214 °C |
MDL Number:
MFCD00664644 CAS Number: 96017-10-6 |
Specification Test Results
Melting point | 212-214°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...