1-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

045599-500MG 045599-1G 045599-5G
101871-688EA 747.69 USD
101871-688 101871-690 101871-692
1-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%
1-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₇ClF₃N₃O₂S
Melting Pt: 195…198 °C
MDL Number: MFCD00665287
CAS Number: 159885-82-2

Order Now

Specification Test Results

Melting point 195-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR