Ethyl-1-(4-phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

045597-500MG 045597-1G 045597-5G
101871-676EA 747.69 USD
101871-676 101871-678 101871-680
Ethyl-1-(4-phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(4-phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₂F₃N₃O₂S
MW: 367.35 g/mol
Melting Pt: 93…95 °C
MDL Number: MFCD00665285
CAS Number: 159885-61-7

Order Now

Specification Test Results

Melting point 93-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR